siRNA Details
| Gene Symbol: 2810403D21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015493.2 | 5 | 861 | |||
| GenBank mRNA | AK086455.1 | - | 5 | 862 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2810403D21Rik| Gene Symbol: 2810403D21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015493.2 | 5 | 861 | |||
| GenBank mRNA | AK086455.1 | - | 5 | 862 | ||