siRNA Details
| Gene Symbol: Zfp389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026798.1 | 2 | 749 | |||
| GenBank mRNA | AK132930.1 | - | 2 | 750 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV266899.1 | - | 1 | 80 | Chr. 13 - 21504315 - 21504580 [-] on Build GRCm38 | |

Zfp389
Genomic Map