siRNA Details
| Gene Symbol: Zfp389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026798.1 | 2 | 399 | |||
| GenBank mRNA | AK132930.1 | - | 2 | 400 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp389| Gene Symbol: Zfp389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026798.1 | 2 | 399 | |||
| GenBank mRNA | AK132930.1 | - | 2 | 400 | ||