siRNA Details
| Gene Symbol: Gnas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003258.1 | 1, 2 | 24 | |||
| GenBank mRNA | AK020895.1 | - | 1, 2 | 209 | ||
| AY682719.1 | - | 1, 2 | 24 | |||
| BC062654.1 | - | 1, 2 | 170 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gnas| Gene Symbol: Gnas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003258.1 | 1, 2 | 24 | |||
| GenBank mRNA | AK020895.1 | - | 1, 2 | 209 | ||
| AY682719.1 | - | 1, 2 | 24 | |||
| BC062654.1 | - | 1, 2 | 170 | |||