siRNA Details
| Gene Symbol: D2Wsu81e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044983.1 | - | 1 | 322 | ||
| BY289626.1 | - | 1 | 340 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D2Wsu81e| Gene Symbol: D2Wsu81e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044983.1 | - | 1 | 322 | ||
| BY289626.1 | - | 1 | 340 | |||