siRNA Details
| Gene Symbol: Copg2os2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002845.1 | 284 | ||||
| GenBank mRNA | U20264.1 | - | 284 | |||
| U20265.1 | - | 1410 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Copg2os2| Gene Symbol: Copg2os2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002845.1 | 284 | ||||
| GenBank mRNA | U20264.1 | - | 284 | |||
| U20265.1 | - | 1410 | ||||