siRNA Details
| Gene Symbol: Ythdf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027375.1 | 3 | 455 | |||
| GenBank mRNA | AK039176.1 | - | 3 | 402 | ||
| AK053736.1 | - | 3 | 374 | |||
| BC057158.1 | - | 3 | 366 | 5348717 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ythdf3| Gene Symbol: Ythdf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027375.1 | 3 | 455 | |||
| GenBank mRNA | AK039176.1 | - | 3 | 402 | ||
| AK053736.1 | - | 3 | 374 | |||
| BC057158.1 | - | 3 | 366 | 5348717 | ||