siRNA Details
| Gene Symbol: Ythdf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027375.1 | 3, 4 | 505 | |||
| GenBank mRNA | AK039176.1 | - | 3, 4 | 452 | ||
| AK053736.1 | - | 3, 4 | 424 | |||
| BC057158.1 | - | 3, 4 | 416 | 5348717 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ythdf3| Gene Symbol: Ythdf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027375.1 | 3, 4 | 505 | |||
| GenBank mRNA | AK039176.1 | - | 3, 4 | 452 | ||
| AK053736.1 | - | 3, 4 | 424 | |||
| BC057158.1 | - | 3, 4 | 416 | 5348717 | ||