siRNA Details
Gene Symbol: Ythdf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027375.1 | 3 | 477 | |||
GenBank mRNA | AK039176.1 | - | 3 | 424 | ||
AK053736.1 | - | 3 | 396 | |||
BC057158.1 | - | 3 | 388 | 5348717 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ythdf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027375.1 | 3 | 477 | |||
GenBank mRNA | AK039176.1 | - | 3 | 424 | ||
AK053736.1 | - | 3 | 396 | |||
BC057158.1 | - | 3 | 388 | 5348717 |