siRNA Details
| Gene Symbol: 1700028J19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029436.1 | 5 | 1008 | |||
| GenBank mRNA | AK006462.1 | - | 5 | 1070 | ||
| BC049653.1 | - | 5 | 974 | |||
| BC061239.1 | - | 4 | 884 | |||
| BC087938.1 | - | 4 | 884 | |||
| BC100477.1 | - | 5 | 990 | |||

1700028J19Rik
Genomic Map