siRNA Details
| Gene Symbol: 1700028J19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029436.1 | 2 | 137 | |||
| GenBank mRNA | AK006462.1 | - | 2 | 138 | ||
| BC049653.1 | - | 2 | 103 | |||
| BC061239.1 | - | 1 | 18 | |||
| BC087938.1 | - | 1 | 18 | |||
| BC100477.1 | - | 2 | 119 | |||

1700028J19Rik
Genomic Map