siRNA Details
| Gene Symbol: E130114P18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015513.2 | 3 | 1053 | |||
| GenBank mRNA | AK082601.1 | - | 3 | 1055 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E130114P18Rik| Gene Symbol: E130114P18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015513.2 | 3 | 1053 | |||
| GenBank mRNA | AK082601.1 | - | 3 | 1055 | ||