siRNA Details
| Gene Symbol: 9330151L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033222.1 | 2 | 2461 | |||
| GenBank mRNA | AK034052.1 | - | 2 | 2449 | ||
| AK154174.1 | - | 2 | 2461 | |||
| AK161156.1 | - | 1 | 2578 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9330151L19Rik| Gene Symbol: 9330151L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033222.1 | 2 | 2461 | |||
| GenBank mRNA | AK034052.1 | - | 2 | 2449 | ||
| AK154174.1 | - | 2 | 2461 | |||
| AK161156.1 | - | 1 | 2578 | |||