siRNA Details
| Gene Symbol: Snord22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_004445.1 | 1 | 58 | |||
| Gene Symbol: Snhg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036616.1 | - | 2 | 2210 | ||
| AK051045.1 | - | 3 | 1966 | |||

Snord22
Genomic Map