siRNA Details
| Gene Symbol: 4732416N19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015615.1 | 2 | 533 | |||
| GenBank mRNA | AK005757.1 | - | 2 | 533 | ||
| AK033816.1 | - | 692 | ||||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY705917.1 | - | 2 | 533 | Chr. 6 - 148212288 - 148235666 [+] on Build GRCm38 | |

4732416N19Rik
Genomic Map