siRNA Details
| Gene Symbol: A830082N09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015526.2 | 1 | 6517 | |||
| GenBank mRNA | AK044037.1 | - | 2 | 301 | ||
| AK136944.1 | - | 1 | 1537 | |||
| BE653894.1 | - | 1 | 271 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A830082N09Rik| Gene Symbol: A830082N09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015526.2 | 1 | 6517 | |||
| GenBank mRNA | AK044037.1 | - | 2 | 301 | ||
| AK136944.1 | - | 1 | 1537 | |||
| BE653894.1 | - | 1 | 271 | |||