siRNA Details
| Gene Symbol: Gm10012 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028042.1 | 1 | 235 | |||
| GenBank mRNA | CA533111.1 | - | 1 | 235 | ||
| Gene Symbol: Gm12338 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK137335.1 | - | 1496 | |||

Gm10012
Genomic Map