siRNA Details
| Gene Symbol: Dancr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015531.1 | 2 | 951 | |||
| GenBank mRNA | AK012278.1 | - | 3 | 448 | ||
| AK045300.1 | - | 1192 | ||||
| AK172090.1 | - | 2 | 951 | |||
| AK172703.1 | - | 1 | 1134 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dancr| Gene Symbol: Dancr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015531.1 | 2 | 951 | |||
| GenBank mRNA | AK012278.1 | - | 3 | 448 | ||
| AK045300.1 | - | 1192 | ||||
| AK172090.1 | - | 2 | 951 | |||
| AK172703.1 | - | 1 | 1134 | |||