siRNA Details
| Gene Symbol: 1700049L16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003644.1 | 1 | 623 | |||
| GenBank mRNA | AK075762.1 | - | 1 | 628 | ||
| BC048504.1 | - | 1 | 623 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV045951.3 | - | 1 | 820 | Chr. 10 - 71979886 - 71980698 [-] on Build GRCm38 | |

1700049L16Rik
Genomic Map