siRNA Details
| Gene Symbol: Sec14l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029459.1 | 3 | 676 | |||
| GenBank mRNA | BC055849.1 | - | 3 | 596 | ||
| CJ066501.1 | - | 3 | 613 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sec14l1| Gene Symbol: Sec14l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029459.1 | 3 | 676 | |||
| GenBank mRNA | BC055849.1 | - | 3 | 596 | ||
| CJ066501.1 | - | 3 | 613 | |||