siRNA Details
| Gene Symbol: 2010320M18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029440.1 | 1 | 693 | |||
| GenBank mRNA | AK008596.1 | - | 1 | 694 | ||
| AK162274.1 | - | 1 | 633 | |||
| BC051496.1 | - | 1 | 657 | |||
| BC059891.1 | - | 1 | 611 | |||
| BC092380.1 | - | 1 | 617 | |||

2010320M18Rik
Genomic Map