siRNA Details
| Gene Symbol: Snhg7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024068.2 | 4 | 434 | |||
| GenBank mRNA | AK011283.1 | - | 4 | 435 | ||
| AK028166.1 | - | 4 | 435 | |||
| BC046478.1 | - | 3 | 2153 | |||
| CB245554.1 | - | 1 | 142 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snhg7| Gene Symbol: Snhg7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024068.2 | 4 | 434 | |||
| GenBank mRNA | AK011283.1 | - | 4 | 435 | ||
| AK028166.1 | - | 4 | 435 | |||
| BC046478.1 | - | 3 | 2153 | |||
| CB245554.1 | - | 1 | 142 | |||