siRNA Details
| Gene Symbol: Snhg7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024068.2 | 4 | 729 | |||
| GenBank mRNA | AK028166.1 | - | 4 | 730 | ||
| CB245554.1 | - | 1 | 437 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY642400.1 | - | 3 | 392 | Chr. 2 - 26637177 - 26639476 [-] on Build GRCm38 | |

Snhg7
Genomic Map