siRNA Details
| Gene Symbol: Gm6524 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028307.1 | 1 | 43 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm6524| Gene Symbol: Gm6524 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028307.1 | 1 | 43 | |||