siRNA Details
| Gene Symbol: C130026L21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015546.2 | 1, 2 | 321 | |||
| GenBank mRNA | AK160717.1 | - | 1, 2 | 138 | ||
| AK160721.1 | - | 1, 2 | 322 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C130026L21Rik| Gene Symbol: C130026L21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015546.2 | 1, 2 | 321 | |||
| GenBank mRNA | AK160717.1 | - | 1, 2 | 138 | ||
| AK160721.1 | - | 1, 2 | 322 | |||