siRNA Details
| Gene Symbol: 6530402F18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029460.1 | 2 | 4203 | |||
| GenBank mRNA | AK079298.1 | - | 2 | 4198 | ||
| AK220426.1 | - | 3479 | ||||
| BC066084.1 | - | 2 | 4203 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

6530402F18Rik| Gene Symbol: 6530402F18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029460.1 | 2 | 4203 | |||
| GenBank mRNA | AK079298.1 | - | 2 | 4198 | ||
| AK220426.1 | - | 3479 | ||||
| BC066084.1 | - | 2 | 4203 | |||