siRNA Details
| Gene Symbol: Rab10os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015551.1 | 7 | 731 | |||
| GenBank mRNA | AK007702.1 | - | 7 | 732 | ||
| BC080728.1 | - | 7 | 2528 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab10os| Gene Symbol: Rab10os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015551.1 | 7 | 731 | |||
| GenBank mRNA | AK007702.1 | - | 7 | 732 | ||
| BC080728.1 | - | 7 | 2528 | |||