siRNA Details
| Gene Symbol: Rny1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_004419.1 | 1 | 63 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | U84681.1 | - | 1 | 46 | Chr. - - [-] on Build | |

Rny1
Genomic Map