siRNA Details
| Gene Symbol: Vmn2r29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003555.1 | 7 | 992 | |||
| GenBank mRNA | AK039785.1 | - | 1 | 2855 | ||
| BC096375.1 | - | 7 | 918 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn2r29| Gene Symbol: Vmn2r29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003555.1 | 7 | 992 | |||
| GenBank mRNA | AK039785.1 | - | 1 | 2855 | ||
| BC096375.1 | - | 7 | 918 | |||