siRNA Details
| Gene Symbol: Gm8979 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030719.1 | 1 | 802 | |||
| GenBank mRNA | BC030868.1 | - | 1 | 802 | ||
| Gene Symbol: Gm8995 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK134184.1 | - | 1 | 1750 | ||

Gm8979
Genomic Map