siRNA Details
| Gene Symbol: Rhno1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027359.1 | 3 | 1132 | |||
| NR_027360.1 | 3 | 1528 | ||||
| GenBank mRNA | AK020031.1 | - | 3 | 1382 | ||
| AK156717.1 | - | 4 | 1646 | |||
| AK168313.1 | - | 1575 | ||||
| BC027368.1 | - | 3 | 1530 | |||

Rhno1
Genomic Map