siRNA Details
| Gene Symbol: Rhno1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027359.1 | 3 | 949 | |||
| NR_027360.1 | 3 | 1345 | ||||
| GenBank mRNA | AK011167.1 | - | 3 | 1142 | ||
| AK020031.1 | - | 3 | 1199 | |||
| AK029454.1 | - | 3 | 1031 | |||
| AK146167.1 | - | 3 | 920 | |||
| AK156717.1 | - | 4 | 1463 | |||
| AK168313.1 | - | 1392 | ||||
| BC005726.1 | - | 1 | 566 | |||
| BC027368.1 | - | 3 | 1347 | |||

Rhno1
Genomic Map