siRNA Details
| Gene Symbol: Rhno1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027359.1 | 3 | 341 | |||
| NR_027360.1 | 3 | 737 | ||||
| GenBank mRNA | AK011109.1 | - | 3 | 433 | ||
| AK011167.1 | - | 3 | 535 | |||
| AK020031.1 | - | 3 | 590 | |||
| AK029454.1 | - | 3 | 423 | |||
| AK146167.1 | - | 3 | 312 | |||
| AK156717.1 | - | 4 | 855 | |||
| AK168313.1 | - | 784 | ||||
| BC027368.1 | - | 3 | 739 | |||
| BE655013.1 | - | 3 | 341 | |||

Rhno1
Genomic Map