siRNA Details
Gene Symbol: Rhno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027359.1 | 3 | 1198 | |||
NR_027360.1 | 3 | 1594 | ||||
GenBank mRNA | AK156717.1 | - | 4 | 1712 | ||
AK168313.1 | - | 1641 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rhno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027359.1 | 3 | 1198 | |||
NR_027360.1 | 3 | 1594 | ||||
GenBank mRNA | AK156717.1 | - | 4 | 1712 | ||
AK168313.1 | - | 1641 |