siRNA Details
| Gene Symbol: Rhno1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027359.1 | 3 | 1244 | |||
| NR_027360.1 | 3 | 1640 | ||||
| GenBank mRNA | AK156717.1 | - | 4 | 1758 | ||
| AK168313.1 | - | 1687 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhno1| Gene Symbol: Rhno1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027359.1 | 3 | 1244 | |||
| NR_027360.1 | 3 | 1640 | ||||
| GenBank mRNA | AK156717.1 | - | 4 | 1758 | ||
| AK168313.1 | - | 1687 | ||||