siRNA Details
| Gene Symbol: Gm10069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028592.1 | 1 | 213 | |||
| NR_028593.1 | 1 | 213 | ||||
| GenBank mRNA | AK046088.1 | - | 1 | 169 | ||
| AK086937.1 | - | 1 | 214 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10069| Gene Symbol: Gm10069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028592.1 | 1 | 213 | |||
| NR_028593.1 | 1 | 213 | ||||
| GenBank mRNA | AK046088.1 | - | 1 | 169 | ||
| AK086937.1 | - | 1 | 214 | |||