siRNA Details
| Gene Symbol: Gm10069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028592.1 | 2 | 725 | |||
| NR_028593.1 | 2 | 725 | ||||
| GenBank mRNA | AK029950.1 | - | 2 | 427 | ||
| AK046088.1 | - | 2 | 681 | |||
| AK086937.1 | - | 2 | 726 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10069| Gene Symbol: Gm10069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028592.1 | 2 | 725 | |||
| NR_028593.1 | 2 | 725 | ||||
| GenBank mRNA | AK029950.1 | - | 2 | 427 | ||
| AK046088.1 | - | 2 | 681 | |||
| AK086937.1 | - | 2 | 726 | |||