siRNA Details
| Gene Symbol: Gm10069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028592.1 | 2 | 647 | |||
| NR_028593.1 | 2 | 647 | ||||
| GenBank mRNA | AK029950.1 | - | 2 | 349 | ||
| AK046088.1 | - | 2 | 603 | |||
| AK086937.1 | - | 2 | 648 | |||
| BC091771.1 | - | 2 | 65 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG970567.1 | - | 2 | 65 | Chr. 6 - 128439339 - 128503258 [+] on Build GRCm38 | |

Gm10069
Genomic Map