siRNA Details
| Gene Symbol: A930013F10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027886.1 | 1 | 2233 | |||
| GenBank mRNA | AK020821.1 | - | 1 | 999 | ||
| AK020851.1 | - | 1 | 1001 | |||
| AK052615.1 | - | 1 | 1714 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A930013F10Rik| Gene Symbol: A930013F10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027886.1 | 1 | 2233 | |||
| GenBank mRNA | AK020821.1 | - | 1 | 999 | ||
| AK020851.1 | - | 1 | 1001 | |||
| AK052615.1 | - | 1 | 1714 | |||