siRNA Details
| Gene Symbol: BC064078 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015455.1 | 3 | 489 | |||
| GenBank mRNA | AK134020.1 | - | 452 | |||
| BC016205.1 | - | 3 | 502 | |||
| BC017132.1 | - | 3 | 502 | |||
| BC025095.1 | - | 3 | 486 | |||
| BC038635.1 | - | 3 | 485 | |||
| BC051485.1 | - | 3 | 548 | |||
| BC064078.1 | - | 3 | 478 | |||
| BC096405.1 | - | 3 | 489 | |||

BC064078
Genomic Map