siRNA Details
| Gene Symbol: BC064078 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015455.1 | 1 | 43 | |||
| GenBank mRNA | BC016205.1 | - | 1 | 56 | ||
| BC017132.1 | - | 1 | 56 | |||
| BC038635.1 | - | 1 | 39 | |||
| BC051485.1 | - | 1 | 102 | |||
| BC064078.1 | - | 1 | 32 | |||
| BC096405.1 | - | 1 | 43 | |||

BC064078
Genomic Map