siRNA Details
| Gene Symbol: Gm5434 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003649.1 | 1 | 1284 | |||
| GenBank mRNA | AK136183.1 | - | 1 | 1286 | ||
| BC085248.1 | - | 1 | 1292 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5434| Gene Symbol: Gm5434 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003649.1 | 1 | 1284 | |||
| GenBank mRNA | AK136183.1 | - | 1 | 1286 | ||
| BC085248.1 | - | 1 | 1292 | |||