siRNA Details
| Gene Symbol: Snhg17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015463.1 | 6 | 2205 | |||
| NR_027887.1 | 6 | 1986 | ||||
| GenBank mRNA | AK133696.1 | - | 1 | 2190 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snhg17| Gene Symbol: Snhg17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015463.1 | 6 | 2205 | |||
| NR_027887.1 | 6 | 1986 | ||||
| GenBank mRNA | AK133696.1 | - | 1 | 2190 | ||