siRNA Details
| Gene Symbol: Rnf138rt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006700.1 | - | 1 | 1007 | ||
| BC125311.1 | - | 1 | 936 | |||
| BC125313.1 | - | 1 | 936 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA464092.1 | - | 1 | 682 | Chr. X - 163760160 - 163761004 [-] on Build GRCm38 | |

Rnf138rt1
Genomic Map