siRNA Details
| Gene Symbol: Rnf138rt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006700.1 | - | 1 | 401 | ||
| BC125311.1 | - | 1 | 330 | |||
| BC125313.1 | - | 1 | 330 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf138rt1| Gene Symbol: Rnf138rt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006700.1 | - | 1 | 401 | ||
| BC125311.1 | - | 1 | 330 | |||
| BC125313.1 | - | 1 | 330 | |||