siRNA Details
| Gene Symbol: A330076H08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015599.2 | 1, 2 | 301 | |||
| GenBank mRNA | AK036297.1 | - | 1, 2 | 302 | ||
| AK038438.1 | - | 355 | ||||
| AK039637.1 | - | 1, 2 | 329 | |||
| AK080655.2 | - | 358 | ||||
| AK083195.1 | - | 1, 2 | 354 | |||

A330076H08Rik
Genomic Map