siRNA Details
| Gene Symbol: A330076H08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015599.2 | 4 | 896 | |||
| GenBank mRNA | AK036297.1 | - | 4 | 897 | ||
| AK039637.1 | - | 4 | 924 | |||
| AK083195.1 | - | 4 | 949 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A330076H08Rik| Gene Symbol: A330076H08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015599.2 | 4 | 896 | |||
| GenBank mRNA | AK036297.1 | - | 4 | 897 | ||
| AK039637.1 | - | 4 | 924 | |||
| AK083195.1 | - | 4 | 949 | |||