siRNA Details
| Gene Symbol: Fam120aos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015601.1 | 2 | 1273 | |||
| GenBank mRNA | AK021143.1 | - | 2 | 965 | ||
| BC045201.1 | - | 2 | 1205 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam120aos| Gene Symbol: Fam120aos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015601.1 | 2 | 1273 | |||
| GenBank mRNA | AK021143.1 | - | 2 | 965 | ||
| BC045201.1 | - | 2 | 1205 | |||