siRNA Details
| Gene Symbol: Gm4792 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033209.1 | 1 | 25 | |||
| GenBank mRNA | BC092352.1 | - | 1 | 25 | 5400830 | |
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI872399.1 | - | 1 | 24 | Chr. 10 - 94293667 - 94298702 [-] on Build GRCm38 | |

Gm4792
Genomic Map