siRNA Details
Gene Symbol: Gm4792 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033209.1 | 3 | 735 | |||
GenBank mRNA | AK053374.1 | - | 3 | 689 | ||
BC092352.1 | - | 3 | 735 | 5400830 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm4792 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033209.1 | 3 | 735 | |||
GenBank mRNA | AK053374.1 | - | 3 | 689 | ||
BC092352.1 | - | 3 | 735 | 5400830 |